missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dicloroacetofenona, 98 %, Thermo Scientific Chemicals

Catalog Number 11440543
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
56041-57-7
C8H6Cl2O
189.04
MFCD00052988
KMABBMYSEVZARZ-UHFFFAOYSA-N
1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
91986
1-(2,3-dichlorophenyl)ethan-1-one
CC(=O)C1=CC=CC(Cl)=C1Cl
Este artículo no se puede devolver. Vea la política de devoluciones
56041-57-7
C8H6Cl2O
189.04
MFCD00052988
KMABBMYSEVZARZ-UHFFFAOYSA-N
1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
91986
1-(2,3-dichlorophenyl)ethan-1-one
CC(=O)C1=CC=CC(Cl)=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2',3'-Dichloroacetophenone
Densidad 1.293
Punto de ebullición 106°C to 108°C (2 mmHg)
Índice de refracción 1.56
Cantidad 25 g
Beilstein 2438815
Formula Weight (peso de la fórmula) 189.04
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'