missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de 2-[3,4-(metilendioxi)fenil]etilamina, 95 %, Thermo Scientific Chemicals

Catalog Number p-7040436
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1653-64-1
C9H12ClNO2
201.65
MFCD00035193
NDYXFQODWGEGNU-UHFFFAOYSA-N
3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
74246
2-(1,3-benzodioxol-5-il)etanamina;clorhidrato
C1OC2=C(O1)C=C(C=C2)CCN.Cl
Este artículo no se puede devolver. Vea la política de devoluciones
1653-64-1
C9H12ClNO2
201.65
MFCD00035193
NDYXFQODWGEGNU-UHFFFAOYSA-N
3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
74246
2-(1,3-benzodioxol-5-il)etanamina;clorhidrato
C1OC2=C(O1)C=C(C=C2)CCN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride
Punto de fusión 213°C to 214°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 201.65
Porcentaje de pureza 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'