Learn More
2,3,4,5-Tetrafluorobenzoylacetic Acid Ethyl Ester, TRC
Especificaciones
Especificaciones
| Chemical Name or Material | 2,3,4,5-Tetrafluorobenzoylacetic Acid Ethyl Ester |
| CAS | 94695-50-8 |
| Molecular Formula | C11H8F4O3 |
| Quantity | 10 g |
| Synonym | 2,3,4,5-tetrafluoro-β-oxobenzenepropanoic Acid Ethyl Ester, Ethyl 2,3,4,5-Tetrafluoro-β-oxobenzenepropanoate, Ethyl 2,3,4,5-tetrafluoro-β-oxobenzenepropionate, Ethyl 2,3,4,5-Tetrafluorobenzoylacetate, Ethyl 2-(2,3,4,5-Tetrafluorobenzoyl)acetate, Ethyl α-(2,3,4,5-Tetrafluorobenzoyl)acetate |
| InChI Formula | InChI=1S/C11H8F4O3/c1-2-18-8(17)4-7(16)5-3-6(12)10(14)11(15)9(5)13/h3H,2,4H2,1H3 |
| SMILES | CCOC(=O)CC(=O)c1cc(F)c(F)c(F)c1F |
| IUPAC Name | ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate |
| Molecular Weight (g/mol) | 264.17 |
| Formula Weight | 264.041 |