missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naftoxi)etanol, + 98 %, Thermo Scientific Chemicals

Catalog Number p-7081553
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
93-20-9
C12H12O2
188.226
MFCD00016809
BQPBZDSDFCDSAO-UHFFFAOYSA-N
2-2-naphthyloxy ethanol, 2-2-naphthoxy ethanol, 2-naphthalen-2-yloxy ethanol, anavenol, 2-2-hydroxyethoxy naphthalene, ethanol, 2-2-naphthalenyloxy, 2-2-naphthalenyloxy ethanol, unii-7m7cut7ccu, ethylene glycol mono-2-naphthyl ether, .beta.-naphthoxyethanol
7131
2-naftaleno-2-iloxietanol
C1=CC=C2C=C(C=CC2=C1)OCCO
Este artículo no se puede devolver. Vea la política de devoluciones
93-20-9
C12H12O2
188.226
MFCD00016809
BQPBZDSDFCDSAO-UHFFFAOYSA-N
2-2-naphthyloxy ethanol, 2-2-naphthoxy ethanol, 2-naphthalen-2-yloxy ethanol, anavenol, 2-2-hydroxyethoxy naphthalene, ethanol, 2-2-naphthalenyloxy, 2-2-naphthalenyloxy ethanol, unii-7m7cut7ccu, ethylene glycol mono-2-naphthyl ether, .beta.-naphthoxyethanol
7131
2-naftaleno-2-iloxietanol
C1=CC=C2C=C(C=CC2=C1)OCCO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2- (2-Naphthoxy)ethanol
Punto de fusión 72°C to 76°C
Cantidad 5 g
Beilstein 2086973
Índice Merck 14,6413
Formula Weight (peso de la fórmula) 188.23
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'