Learn More
2-(2-Methoxyphenoxy)ethylamine Hydrochloride, Mikromol™
Product Code. 30050975
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050975 25 mg 25mg
30049414 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050975 Supplier Mikromol Supplier No. MM0291.110025
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(2-Methoxyphenoxy)ethylamine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine Hydrochloride
CAS 64464-07-9
Analyte or Component Names 2-(2-Methoxyphenoxy)ethylamine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H13 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym 2-(2-Methoxyphenoxy)-ethanamine hydrochloride, 2-(o-Methoxyphenoxy)-ethylamine hydrochloride, 2-(2-Methoxyphenoxy)ethylamine Hydrochloride, Carvedilol Related Compound E as Hydrochloride
InChI Formula InChI=1S/C9H13NO2.ClH/c1-11-8-4-2-3-5-9(8)12-7-6-10;/h2-5H,6-7,10H2,1H3;1H
SMILES Cl.COc1ccccc1OCCN
IUPAC Name 2-(2-methoxyphenoxy)ethanamine;hydrochloride
Molecular Weight (g/mol) 203.67
Quantity 25 mg
Formula Weight 203.0713 g/mol