Learn More
2,2,2-Trifluoro-N-[2-(4-iodo-2,5-dimethoxyphenyl)ethyl]acetamide, TRC
Product Code. 30456956
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30456956 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 30456956 Supplier Toronto Research Chemicals Supplier No. TRCT78306550MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2,2-Trifluoro-N-[2-(4-iodo-2,5-dimethoxyphenyl)ethyl]acetamide
CAS 159277-10-8
Molecular Formula C12H13F3INO3
Quantity 50 mg
InChI Formula InChI=1S/C12H13F3INO3/c1-19-9-6-8(16)10(20-2)5-7(9)3-4-17-11(18)12(13,14)15/h5-6H,3-4H2,1-2H3,(H,17,18)
SMILES COc1cc(CCNC(=O)C(F)(F)F)c(OC)cc1I
IUPAC Name 2,2,2-trifluoro-N-[2-(4-iodo-2,5-dimethoxyphenyl)ethyl]acetamide
Molecular Weight (g/mol) 403.14
Formula Weight 402.9892