Learn More
Bochem™ 18/10 Steel Shells
Product Code. 10596255
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
10596255 4 L Each
1 options
This item is not returnable. View return policy
Product Code. 10596255 Supplier Bochem™ Supplier No. 8606
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Material Stainless Steel (18/10)
Capacity (Metric) 4 L
Height (Metric) 135 mm
Diameter (Metric) 245 mm
Shape High
Product Type Shells