Learn More
Applied Biosystems™ 16-Capillary Array For 3130xl/3100 Genetic Analyzer
Use for fragment analysis and rapid sequencing applications.
Marca: Applied Biosystems™ 4319899
Detalles adicionales : Peso : 0.01000kg
Descripción
- For the 3130xl and 3100 Genetic Analyzer
- Usage per array is 100 runs (with use of Applied Biosystems reagents).
Order Info
Each
Especificación
16-Capillary Array, 80cm Capillary Length, 91cm x 50μm I.D. Capillary, 170 min. Run Time, Use for Short Fragment Analysis, Room Temperature, For Use With 3130xl/3100 Genetic Analyzer | |
3100 Genetic Analyzer, 3130xl Genetic Analyzer | |
1 Array | |
170 min. | |
100 Runs |
Fragment Analysis, Sequencing | |
16-Capillary Array | |
Ambient Temperature | |
1 Array |
For research use only