missing translation for 'onlineSavingsMsg'
Learn More

12-Chloro-5-dodecyne, 98%, Thermo Scientific™

Product Code. 11405596
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11405596 5 g 5g
11415596 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11405596

Brand: Thermo Scientific Alfa Aesar B20730.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42513-36-0
Molecular Formula C12H21Cl
Molecular Weight (g/mol) 200.75
MDL Number MFCD00464356
InChI Key DJJGBABLSKSRGV-UHFFFAOYSA-N
Synonym 12-chloro-5-dodecyne, 5-dodecyne, 12-chloro, 1-chloro-7-dodecyne, 12-chloranyldodec-5-yne, 12-chloro-dodec-5-yne, acmc-1ao5f, n-butyl 6-chlorohexyl acetylene
PubChem CID 123509
IUPAC Name 12-chlorododec-5-yne
SMILES CCCCC#CCCCCCCCl

Specifications

Quantity 5 g
Formula Weight 198.74
Percent Purity 98%
Chemical Name or Material 12-Chloro-5-dodecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.