Learn More
(11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic Acid (Isoxepac), Mikromol™
Product Code. 30049314
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
30049314 100 mg 250mg
1 options
This item is not returnable. View return policy
Product Code. 30049314 Supplier Mikromol Supplier No. MM0430.00
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic Acid (Isoxepac)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (11-oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid
CAS 55453-87-7
Analyte or Component Names (11-oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid
Shipping Condition Room Temperature
Molecular Formula C16 H12 O4
Certifications/Compliance ISO 17034
Synonym 6,11-Dihydro-11-oxodibenz[b,e]oxepin-2-acetic acid, 11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-acetic acid, 11-Oxo-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid, 11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-acetic acid, 6,11-Dihydro-11-oxodibenz[b,e]oxepin-2-acetic acid, Artil, HP 549, Isoxepac, NSC 300907, P 720549, Dibenz[b,e]oxepin-2-acetic acid, 6,11-dihydro-11-oxo-, Olopatadine Related Compound C
InChI Formula InChI=1S/C16H12O4/c17-15(18)8-10-5-6-14-13(7-10)16(19)12-4-2-1-3-11(12)9-20-14/h1-7H,8-9H2,(H,17,18)
SMILES OC(=O)Cc1ccc2OCc3ccccc3C(=O)c2c1
IUPAC Name 2-(11-oxo-6H-benzo[c][1]benzoxepin-2-yl)acetic acid
Molecular Weight (g/mol) 268.26
Quantity 100 mg
Formula Weight 268.0736 g/mol