Learn More
11,12-Didehydro Retinol, TRC
Product Code. 30375419
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
30375419 1 mg 1mg
1 options
This item is not returnable. View return policy
Product Code. 30375419 Supplier Toronto Research Chemicals Supplier No. TRCD4394101MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 11,12-Didehydro Retinol
CAS 29443-88-7
Molecular Formula C20 H28 O
Quantity 1 mg
Synonym 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,6,8-nonatrien-4-yn-1-ol, 11,12-Didehydroretinol
InChI Formula InChI=1S/C20H28O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h8,11-13,21H,7,10,14-15H2,1-5H3/b12-11+,16-8+,17-13+
SMILES C\C(=C/C#C\C(=C\CO)\C)\C=C\C1=C(C)CCCC1(C)C
IUPAC Name (2E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,6,8-trien-4-yn-1-ol
Molecular Weight (g/mol) 284.436
Formula Weight 284.214