Learn More
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC
Product Code. 30380229
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30380229 10 mg 10mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380229 Supplier Toronto Research Chemicals Supplier No. TRCD44913710MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10,11-Dihydro-10-hydroxycarbazepine-D4 (Major)
CAS 1020719-39-4
Purity Grade Notes HPLC
Molecular Formula C15 2H4 H10 N2 O2
Quantity 10 mg
Synonym Licarbazepine D4
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)/i1D,3D,5D,7D
SMILES [2H]c1c([2H])c([2H])c2N(C(=O)N)c3ccccc3C(O)Cc2c1[2H]
IUPAC Name 1,2,3,4-tetradeuterio-6-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Molecular Weight (g/mol) 258.31
Formula Weight 258.1306
Percent Purity >95