Learn More
1-Tetracosene, TRC
Especificaciones
Especificaciones
| Chemical Name or Material | 1-Tetracosene |
| CAS | 10192-32-2 |
| Molecular Formula | C24 H48 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C24H48/c1-3-5-7-9-11-13-15-17-19-21-23-24-22-20-18-16-14-12-10-8-6-4-2/h3H,1,4-24H2,2H3 |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCC=C |
| IUPAC Name | tetracos-1-ene |
| Molecular Weight (g/mol) | 336.638 |
| Formula Weight | 336.376 |