missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol del ácido 1-fenilsulfonil-1H-pirazol-4-borónico, 95 %, Thermo Scientific™

Catalog Number p-7029487
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1073372-04-9
C15H19BN2O4S
334.197
MFCD09037504
NPRHNPBHUVCHKP-UHFFFAOYSA-N
1-phenylsulfonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-phenylsulfonyl pyrazole-4-boronic acid, pinacol ester, 1-phenylsulfonyl-1h-pyrazole-4-boronic acid pinacol ester, 1-benzenesulfonyl-1h-pyrazole-4-boronic acid pinacol ester, 1-benzenesulfonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzenesulfonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, pubchem18434, 1-phenylsulfonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-benzenesulfonyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole
44755203
1-(bencenosulfonil)-4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)pirazol
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)S(=O)(=O)C3=CC=CC=C3
Este artículo no se puede devolver. Vea la política de devoluciones
1073372-04-9
C15H19BN2O4S
334.197
MFCD09037504
NPRHNPBHUVCHKP-UHFFFAOYSA-N
1-phenylsulfonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-phenylsulfonyl pyrazole-4-boronic acid, pinacol ester, 1-phenylsulfonyl-1h-pyrazole-4-boronic acid pinacol ester, 1-benzenesulfonyl-1h-pyrazole-4-boronic acid pinacol ester, 1-benzenesulfonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzenesulfonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, pubchem18434, 1-phenylsulfonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-benzenesulfonyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole
44755203
1-(bencenosulfonil)-4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)pirazol
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)S(=O)(=O)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Phenylsulfonyl-1H-pyrazole-4-boronic acid pinacol ester
Punto de fusión 125°C to 130°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 334.2
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'