missing translation for 'onlineSavingsMsg'
Learn More

1-Metil-4-(4-nitrofenil)piperazina, 97 %, Thermo Scientific™

Catalog Number 15480469
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
16155-03-6
C11H15N3O2
221.26
MFCD00101003
GZNDUKANJZIZOT-UHFFFAOYSA-N
1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
2825198
1-metil-4-(4-nitrofenil)piperazina
CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
16155-03-6
C11H15N3O2
221.26
MFCD00101003
GZNDUKANJZIZOT-UHFFFAOYSA-N
1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
2825198
1-metil-4-(4-nitrofenil)piperazina
CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Methyl-4-(4-nitrophenyl)piperazine
Cantidad 1 g
Formula Weight (peso de la fórmula) 221.26
Porcentaje de pureza 97%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'