missing translation for 'onlineSavingsMsg'
Learn More

1-Cianometil-2-(3-cianofenil)-4,5-dicianoimidazol, 97 %, Thermo Scientific™

Catalog Number p-7035810
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1347815-42-2
C14H6N6
258.244
MFCD20265160
KQHKTLGFDJPVEP-UHFFFAOYSA-N
1-cyanomethyl-2-3-cyanophenyl-4,5-dicyanoimidazole, 1-cyanomethyl-2-3-cyanophenyl imidazole-4,5-dicarbonitrile
73995593
1-(cianometil)-2-(3-cianofenil)imidazol-4,5-dicarbonitrilo
C1=CC(=CC(=C1)C2=NC(=C(N2CC#N)C#N)C#N)C#N
Este artículo no se puede devolver. Vea la política de devoluciones
1347815-42-2
C14H6N6
258.244
MFCD20265160
KQHKTLGFDJPVEP-UHFFFAOYSA-N
1-cyanomethyl-2-3-cyanophenyl-4,5-dicyanoimidazole, 1-cyanomethyl-2-3-cyanophenyl imidazole-4,5-dicarbonitrile
73995593
1-(cianometil)-2-(3-cianofenil)imidazol-4,5-dicarbonitrilo
C1=CC(=CC(=C1)C2=NC(=C(N2CC#N)C#N)C#N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole
Cantidad 250 mg
Número UN UN3439
Formula Weight (peso de la fórmula) 258.24
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'