Learn More
1-Butylpyrrolidin-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Butylpyrrolidin-2-one |
| CAS | 3470-98-2 |
| Molecular Formula | C8H15NO |
| Quantity | 50 mg |
| Synonym | 1-Butyl-2-pyrrolidinone (ACI), 1-Butyl-2-pyrrolidone, Genagen NBP, N-Butyl-2-pyrrolidinone, N-Butyl-2-pyrrolidone, N-Butylbutyrolactam, N-Butylpyrrolidinone, N-Butylpyrrolidone, 2-Pyrrolidinone, 1-butyl- (6CI, 7CI, 8CI, 9CI, ACI) |
| InChI Formula | InChI=1S/C8H15NO/c1-2-3-6-9-7-4-5-8(9)10/h2-7H2,1H3 |
| SMILES | CCCCN1CCCC1=O |
| IUPAC Name | 1-butylpyrrolidin-2-one |
| Molecular Weight (g/mol) | 141.21 |
| Formula Weight | 141.1154 |