missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(difluorometoxi)benceno, +97 %, Thermo Scientific™

Catalog Number p-7026885
Click to view available options
Cantidad:
2 g
10 g
Este artículo no se puede devolver. Vea la política de devoluciones
175278-33-8
C7H5BrF2O
223.017
MFCD00221468
VHEICCMNYWNFQX-UHFFFAOYSA-N
1-bromo-2-difluoromethoxy benzene, 2-difluoromethoxy bromobenzene, benzene, 1-bromo-2-difluoromethoxy, 2-bromophenyl difluoromethyl ether, 2-bromo-alpha,alpha-difluoroanisole, 1-difluoromethoxy-2-bromobenzene, acmc-209e9t, ksc493k0n, 1-bromo-2-difluoromethoxy-benzene, attercop-chm at123241
2737004
1-Bromo-2-(difluorometoxi)benceno
C1=CC=C(C(=C1)OC(F)F)Br
Este artículo no se puede devolver. Vea la política de devoluciones
175278-33-8
C7H5BrF2O
223.017
MFCD00221468
VHEICCMNYWNFQX-UHFFFAOYSA-N
1-bromo-2-difluoromethoxy benzene, 2-difluoromethoxy bromobenzene, benzene, 1-bromo-2-difluoromethoxy, 2-bromophenyl difluoromethyl ether, 2-bromo-alpha,alpha-difluoroanisole, 1-difluoromethoxy-2-bromobenzene, acmc-209e9t, ksc493k0n, 1-bromo-2-difluoromethoxy-benzene, attercop-chm at123241
2737004
1-Bromo-2-(difluorometoxi)benceno
C1=CC=C(C(=C1)OC(F)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Bromo-2-(difluoromethoxy)benzene
Punto de ebullición 85°C to 87°C (15 mmHg)
Cantidad 2 g
Formula Weight (peso de la fórmula) 223.02
Porcentaje de pureza ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'