missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pirrolidina-3-carboxamida, 96 %, Thermo Scientific™

Catalog Number p-7033071
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
122684-34-8
C10H18N2O3
214.265
MFCD04116210
NHDGOVOBEZPXMY-UHFFFAOYSA-N
3-aminocarbonyl-1-boc-pyrrolidine, 1-boc-3-carbamoylpyrrolidine, 1-boc-pyrrolidine-3-carboxamide, 3-carbamoyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-pyrrolidine-3-carboxyamide, tert-butyl 3-aminocarbonyl pyrrolidine-1-carboxylate, acmc-20a2cc, ksc496a4t, 1-boc-3-pyrrolidinecarboxamide, 1-n-boc-pyrrolidine-3-carboxyamide
4571096
3-Carbamoilpirrolidina--carboxilato1-carboxilato de terc-butil
CC(C)(C)OC(=O)N1CCC(C1)C(=O)N
Este artículo no se puede devolver. Vea la política de devoluciones
122684-34-8
C10H18N2O3
214.265
MFCD04116210
NHDGOVOBEZPXMY-UHFFFAOYSA-N
3-aminocarbonyl-1-boc-pyrrolidine, 1-boc-3-carbamoylpyrrolidine, 1-boc-pyrrolidine-3-carboxamide, 3-carbamoyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-pyrrolidine-3-carboxyamide, tert-butyl 3-aminocarbonyl pyrrolidine-1-carboxylate, acmc-20a2cc, ksc496a4t, 1-boc-3-pyrrolidinecarboxamide, 1-n-boc-pyrrolidine-3-carboxyamide
4571096
3-Carbamoilpirrolidina--carboxilato1-carboxilato de terc-butil
CC(C)(C)OC(=O)N1CCC(C1)C(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Boc-pyrrolidine-3-carboxamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 214.27
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'