Learn More
Ácido 1-Boc-D-nipecótico, 97 %, Thermo Scientific Chemicals
Catalog Number p-7034718
Click to view available options
Cantidad:
5 g
25 g
163438-09-3
C11H18NO4
228.27
MFCD02179173
NXILIHONWRXHFA-MRVPVSSYSA-M
r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh
643516
CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O
163438-09-3
C11H18NO4
228.27
MFCD02179173
NXILIHONWRXHFA-MRVPVSSYSA-M
r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh
643516
CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O
1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El ácido 1-Boc-D-nipecótico se utiliza como un intermedio de la síntesis química orgánica.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con los agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Boc-D-nipecotic acid
Cantidad 5 g
Beilstein 5539297
Nombre IUPAC (3R)-1-[(terc-butoxi)carbonil]piperidina-3-carboxilato
Formula Weight (peso de la fórmula) 229.28
Porcentaje de pureza 97%