missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-metoxiazetidina, 95 %, Thermo Scientific™

Catalog Number p-7038528
Click to view available options
Cantidad:
250 mg
500 mg
Este artículo no se puede devolver. Vea la política de devoluciones
429669-07-8
C9H17NO3
187.24
MFCD08689580
ZENIBVFXWKRVMK-UHFFFAOYSA-N
1-boc-3-methoxy azetidine, 1-boc-3-methoxyazetidine, n-boc-3-methoxyazetidine, 1-tert-butoxycarbonyl-3-methoxyazetidine, 1-azetidinecarboxylic acid, 3-methoxy-, 1,1-dimethylethyl ester, 1-n-boc-3-methoxyazetidine, 1-boc-3-methoxy ;azetidine, 1-t-butoxycarbonyl-3-methoxyazetidine, tert-butyl3-methoxyazetidine-1-carboxylate, 3-methoxyazetidine, n-boc protected
22243050
tert-butilo 3-metoxiazetidina-1-carboxilato
COC1CN(C1)C(=O)OC(C)(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
429669-07-8
C9H17NO3
187.24
MFCD08689580
ZENIBVFXWKRVMK-UHFFFAOYSA-N
1-boc-3-methoxy azetidine, 1-boc-3-methoxyazetidine, n-boc-3-methoxyazetidine, 1-tert-butoxycarbonyl-3-methoxyazetidine, 1-azetidinecarboxylic acid, 3-methoxy-, 1,1-dimethylethyl ester, 1-n-boc-3-methoxyazetidine, 1-boc-3-methoxy ;azetidine, 1-t-butoxycarbonyl-3-methoxyazetidine, tert-butyl3-methoxyazetidine-1-carboxylate, 3-methoxyazetidine, n-boc protected
22243050
tert-butilo 3-metoxiazetidina-1-carboxilato
COC1CN(C1)C(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-Boc-3-methoxyazetidine
Formula Weight (peso de la fórmula) 187.24
Porcentaje de pureza 95%
Cantidad 250 mg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'