Learn More
1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione, Mikromol™
Product Code. 17953616
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
17953616 25 mg 25mg
1 options
This item is not returnable. View return policy
Product Code. 17953616 Supplier Mikromol Supplier No. MM0344.100025
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione
CAS 720-94-5
Analyte or Component Names 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione
Shipping Condition Room Temperature
Molecular Formula C11 H9 F3 O2
Certifications/Compliance ISO 9001
Synonym 1,3-Butanedione, 4,4,4-trifluoro-1-(4-methylphenyl)-, 1,3-Butanedione, 4,4,4-trifluoro-1-p-tolyl- (6CI,8CI), 4,4,4-Trifluoro-1-(4-methylphenyl)-1,3-butanedione, (4-Methylbenzoyl)trifluoroacetone, 1,1,1-Trifluoro-4-(4-methylphenyl)-2,4-butanedione, 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione, 4-(4-Methylphenyl)-1,1,1-trifluorobutane-2,4-dione, 4-Methyl-1-(4,4,4-trifluoro-3-oxobutanoyl)benzene
InChI Formula InChI=1S/C11H9F3O2/c1-7-2-4-8(5-3-7)9(15)6-10(16)11(12,13)14/h2-5H,6H2,1H3
SMILES Cc1ccc(cc1)C(=O)CC(=O)C(F)(F)F
IUPAC Name 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione
Molecular Weight (g/mol) 230.18
Quantity 25 mg
Formula Weight 230.0555 g/mol