Learn More
1,4-Diisopropylcyclohexane, cis + trans, 98%, Thermo Scientific™
Product Code. 11447194
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447194 25 g 25g
11437194 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447194 Supplier Thermo Scientific Alfa Aesar Supplier No. B21587.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22907-72-8
Molecular Formula C12H24
Molecular Weight (g/mol) 168.324
MDL Number MFCD00019366
InChI Key GMUJGUXGVJZHSK-UHFFFAOYSA-N
Synonym 1,4-diisopropylcyclohexane, 1,4-diisopropyl cyclohexane, 1,4-di-isopropylcyclohexane, 1,4-di propan-2-yl cyclohexane, cyclohexane,1,4-bis 1-methylethyl-,trans, acmc-1cbxy, 1,4-diisopr opylcyclohexane, 1,4-bis methylethyl cyclohexane, 1alpha,4beta-diisopropylcyclohexane, 1,4-diisopropylcyclohexane,cis+trans
PubChem CID 140972
IUPAC Name 1,4-di(propan-2-yl)cyclohexane
SMILES CC(C)C1CCC(CC1)C(C)C
Specifications
Boiling Point 214°C to 216°C
Quantity 25 g
UN Number UN1993
Formula Weight 168.32
Percent Purity 98%
Chemical Name or Material 1,4-Diisopropylcyclohexane, cis + trans