Learn More
1,4-Dihydro-3-nitro-4-oxo-imidazo[5,1-c][1,2,4]triazine-8-carboxamide, TRC
Product Code. 30357382
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
30357382 1 mg 1mg
1 options
This item is not returnable. View return policy
Product Code. 30357382 Supplier Toronto Research Chemicals Supplier No. TRCC9880581MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,4-Dihydro-3-nitro-4-oxo-imidazo[5,1-c][1,2,4]triazine-8-carboxamide
CAS 287964-59-4
Molecular Formula C6 H4 N6 O4
Quantity 1 mg
Synonym 1,4-Dihydro-3-nitro-4-oxo-imidazo[5,1-c][1,2,4]triazine-8-carboxamide, Imidazo[5,1-c][1,2,4]triazine-8-carboxamide, 1,4-dihydro-3-nitro-4-oxo-
InChI Formula InChI=1S/C6H4N6O4/c7-3(13)2-4-9-10-5(12(15)16)6(14)11(4)1-8-2/h1,9H,(H2,7,13)
SMILES NC(=O)c1ncn2C(=O)C(=NNc12)[N+](=O)[O-]
IUPAC Name 3-nitro-4-oxo-1H-imidazo[5,1-c][1,2,4]triazine-8-carboxamide
Molecular Weight (g/mol) 224.134
Formula Weight 224.029