missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibenciloxibenceno, 98 %, Thermo Scientific™

Catalog Number 11441467
Click to view available options
Cantidad:
10 g
50 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
621-91-0
C20H18O2
290.36
MFCD00016875
DYULYMCXVSRUPB-UHFFFAOYSA-N
1,4-dibenzyloxybenzene, 1,4-bis benzyloxy benzene, hydroquinone dibenzyl ether, benzene, 1,4-bis phenylmethoxy, 1,4-bis phenylmethoxy benzene, p-dibenzyloxybenzene, hydrochinon-dibenzylather, 1,4-di benzyloxy benzene, acmc-209n0i, ksc493o6b
69315
1,4-bis(fenilmetoxi)benceno
C(OC1=CC=C(OCC2=CC=CC=C2)C=C1)C1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
621-91-0
C20H18O2
290.36
MFCD00016875
DYULYMCXVSRUPB-UHFFFAOYSA-N
1,4-dibenzyloxybenzene, 1,4-bis benzyloxy benzene, hydroquinone dibenzyl ether, benzene, 1,4-bis phenylmethoxy, 1,4-bis phenylmethoxy benzene, p-dibenzyloxybenzene, hydrochinon-dibenzylather, 1,4-di benzyloxy benzene, acmc-209n0i, ksc493o6b
69315
1,4-bis(fenilmetoxi)benceno
C(OC1=CC=C(OCC2=CC=CC=C2)C=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1,4-Dibenzyloxybenzene
Punto de fusión 125°C to 128°C
Olor Odorless
Cantidad 10 g
Beilstein 2058196
Formula Weight (peso de la fórmula) 290.36
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'