Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
Marca:  Trajan Scientific and Medical™ 0624242
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15288743
  • 61.45€ / paquete de 2
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
Especificación
Especificación
2/Pk.
0.025 mm
100 m
PEEKsil Tubing 1/32 in. OD x 0.025mm ID x 100mm Length
Videos