Learn More
Di-p-toluenosulfonato de 1,3-propanediol, 98 %, Thermo Scientific Chemicals
Catalog Number p-7025550
Click to view available options
Cantidad:
5 g
25 g
5469-66-9
C17H20O6S2
384.46
MFCD00013392
ODVREDGIWSDQFD-UHFFFAOYSA-N
1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate
230370
4-Metilbencenosulfonato de 3-(4-metilfenil)sulfoniloxpropilo
CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1
5469-66-9
C17H20O6S2
384.46
MFCD00013392
ODVREDGIWSDQFD-UHFFFAOYSA-N
1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate
230370
4-Metilbencenosulfonato de 3-(4-metilfenil)sulfoniloxpropilo
CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Se usa como intermediario farmacéutico.
Notas
Almacenar a temperatura ambiente. Manténgase lejos de los agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1,3-Propanediol di-p-toluenesulfonate
Punto de fusión 90°C to 92°C
Cantidad 5 g
Formula Weight (peso de la fórmula) 384.47
Porcentaje de pureza 98%