missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicloroisoquinolina, 97 %, Thermo Scientific™

Catalog Number p-7040178
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
7742-73-6
C9H5Cl2N
198.046
MFCD00034750
BRGZEQXWZWBPJH-UHFFFAOYSA-N
isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
298625
1,3-dicloroisoquinolina
C1=CC=C2C(=C1)C=C(N=C2Cl)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
7742-73-6
C9H5Cl2N
198.046
MFCD00034750
BRGZEQXWZWBPJH-UHFFFAOYSA-N
isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
298625
1,3-dicloroisoquinolina
C1=CC=C2C(=C1)C=C(N=C2Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1,3-Dichloroisoquinoline
Punto de fusión 121°C to 122°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 198.05
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'