missing translation for 'onlineSavingsMsg'
Learn More

1,2-Octanediol, 97 %, Thermo Scientific Chemicals

Catalog Number p-7044745
Click to view available options
Cantidad:
10 g
50 g
Este artículo no se puede devolver. Vea la política de devoluciones
1117-86-8
C8H18O2
146.23
MFCD00010738
AEIJTFQOBWATKX-UHFFFAOYSA-N
1,2-octanediol, 1,2-dihydroxyoctane, 1,2-octylene glycol, capyryl glycol, racemic 1,2-octanediol, acmc-2099as, dsstox_cid_16646, dsstox_rid_79289
14231
CHEBI:34056
octano-1,2-diol
CCCCCCC(CO)O
Este artículo no se puede devolver. Vea la política de devoluciones
1117-86-8
C8H18O2
146.23
MFCD00010738
AEIJTFQOBWATKX-UHFFFAOYSA-N
1,2-octanediol, 1,2-dihydroxyoctane, 1,2-octylene glycol, capyryl glycol, racemic 1,2-octanediol, acmc-2099as, dsstox_cid_16646, dsstox_rid_79289
14231
CHEBI:34056
octano-1,2-diol
CCCCCCC(CO)O

1,2-Octanediol was used as organic modifier to improve the HPLC separation of organic acids and bases. It was also used in preparation of halohydrin palmitates. It is useful for treating head louse infestation clinically. It is also used in coating materials, slurries, paper mills and water circulation systems for the effective preservation against bacteria and fungi. It is used as an emollient, humectant, and wetting agent in cosmetic and skin care products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubilidad
Soluble en agua (3 g/l) a 20 °C.

Notas
Los materiales incompatibles son los agentes oxidantes. Almacenar en un lugar fresco. Mantener el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1,2-Octanediol
Punto de fusión 28°C to 34°C
Punto de ebullición 131°C to 133°C (10 mmHg)
Temperatura de inflamación 138°C (280°F)
Cantidad 10 g
Beilstein 1719619
Información de solubilidad Soluble in water (3g/L) at 20°C.
Formula Weight (peso de la fórmula) 146.23
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'