Learn More
Abcam 1,2-Dimyristoyl-sn-glycero-3-phosphorylcholine (DMPC), Phosphatidylcholine
Product Code. 32112413 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
32112413 250 mg 250mg
1 options
This item is not returnable. View return policy
Product Code. 32112413 Supplier Abcam Supplier No. ab143946250mg
 Restricted Item
Fecha estimada de envoi: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MW 677.9 Da, Purity >98%. Phosphatidylcholine containing myristoyl groups. For use in lipid bilayer studies and in biological systems.
MW 677.9 Da, Purity >98%. Phosphatidylcholine containing myristoyl groups. For use in lipid bilayer studies and in biological systems.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 250 mg
Regulatory Status RUO