Learn More
1-(2-Cloro-4-nitrofenil)piperazina, 97 %, Thermo Scientific™
Catalog Number p-7054812
Click to view available options
Cantidad:
250 mg
1 g
5 g
114878-60-3
C10H12ClN3O2
241.675
MFCD00640776
JNUWBYPJOBSXDN-UHFFFAOYSA-N
2771331
1-(2-cloro-4-nitrofenil)piperazina
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
114878-60-3
C10H12ClN3O2
241.675
MFCD00640776
JNUWBYPJOBSXDN-UHFFFAOYSA-N
2771331
1-(2-cloro-4-nitrofenil)piperazina
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 1-(2-Chloro-4-nitrophenyl)piperazine
Cantidad 250 mg
Sinónimo 1-2-chloro-4-nitrophenyl piperazine, 2-chloro-4-nitrophenyl piperazine, 1-2-chloro-4-nitrophenyl-piperazine, piperazine, 1-2-chloro-4-nitrophenyl, 1-2-chloro-4-nitro-phenyl-piperazine, piperazine,1-2-chloro-4-nitrophenyl, pubchem9343, acmc-1bvwp, cambridge id 5668126, 1-2-chloro-4-nitro-phenyl piperazine
Formula Weight (peso de la fórmula) 241.67
Porcentaje de pureza 97%
Forma física Sólido