missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Bromoetil)-3-(trifluorometil)benceno, 95 %, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Cantidad:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Nombre del producto químico o material | 1-(2-Bromoethyl)-3-(trifluoromethyl)benzene |
Color | Incolora |
Densidad | 1.495 g/mL |
Punto de ebullición | 92°C to 94°C (12 mmHg) |
Índice de refracción | 1.486 |
Cantidad | 250 mg |
Número UN | UN3082 |
Beilstein | 1950052 |
Formula Weight (peso de la fórmula) | 253.06 |
Porcentaje de pureza | 95% |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'