Learn More
1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene, Mikromol™
Product Code. 30051374
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30051374 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 30051374 Supplier Mikromol Supplier No. MM0002.37
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene
CAS 102120-87-6
Analyte or Component Names 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene
Shipping Condition Room Temperature
Molecular Formula C22 H30
Certifications/Compliance ISO 17025
Synonym 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene, Ibuprofen Imp. R (EP), Ibuprofen Impurity R
InChI Formula InChI=1S/C22H30/c1-16(2)14-19-6-10-21(11-7-19)18(5)22-12-8-20(9-13-22)15-17(3)4/h6-13,16-18H,14-15H2,1-5H3
SMILES CC(C)Cc1ccc(cc1)C(C)c2ccc(CC(C)C)cc2
IUPAC Name 1-(2-methylpropyl)-4-[1-[4-(2-methylpropyl)phenyl]ethyl]benzene
Molecular Weight (g/mol) 294.47
Quantity 100 mg
Formula Weight 294.2348 g/mol