missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrilato de 1,1,1,3,3,3-hexafluoroisopropilo, + 98 %, estabilizado con 50 ppm de 4-metoxifenol, Thermo Scientific Chemicals
Click to view available options
Cantidad:
5 g
25 g
Descripción
1,1,1,3,3,3-Hexafluoroisopropyl acrylate is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | 1,1,1,3,3,3-Hexafluoroisopropyl acrylate |
Color | Incolora |
Punto de ebullición | 81°C to 82°C |
Temperatura de inflamación | 17°C |
Envase | Frasco de vidrio |
Fórmula lineal | H2C=CHCO2CH(CF3)2 |
Cantidad | 5 g |
Número UN | UN3272 |
Gravedad específica | 1.37 |
Formula Weight (peso de la fórmula) | 222.09 |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'