Syringe filters for research & pharma
Links
Save Now - Web Exclusive Deals
Take advantage of our web exclusive prices and place your order today.
Sartorius Minisart™ NML Syringe Filters, Sterile
Merck Millex
Must Have
Complete Your Order
Complete your syringeless and syringe filters with the necessary accessories.
  • Great deals
    Eppendorf™ Polypropylene Graduated Microtubes
    Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances or DNA. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
    Shop Now
  • Great deals
    Thermo Scientific™ National Target All-Plastic Disposable Syringes
    Use these all-plastic disposable syringes to dispense small volumes of liquids, or to pressure filter through our syringe filters.
    Shop Now
  • Great deals
    Centrifuge Tubes
    Our range of glass and plastic Centrifuge Tubes are available in a variety of volume capacities, materials, configurations, RCF ratings, and closure types. They may be sterile or non-sterile and may have graduations indicating fill volume.
    Shop Now
Watch This Video
Selection and Use of Syringe Filters
Acrodisc™ Syringe Filters can be used for a variety of applications, including media filtration, clarification, prefiltration, sample clean-up, virus filtration, protein purification, and mycoplasma reduction.
To find the syringe filter that meets your application needs, consider the sterility requirements, the pore size needed, and the membrane type compatible with your process solution. Make sure to also choose a syringe filter with the necessary sample volume throughput.
Acrodisc™ Syringe Filters feature a prefilter for improved filtration of difficult solutions. They are available with a Supor™ membrane, offering higher fluid flow rates and lower protein binding.
Shop Now
Guides to Help You Choose
Whatman Filter Selection Tool Guide
Whatman Filter Selection Tool Guide
The new Whatman™ filter selection tool guides you through the process of selecting an optimal Whatman filter for your application with ease.
Discover More
Pall Acrodisc Syringe Filters
Pall Acrodisc Syringe Filters
Find the right Acrodisc™ syringe filters suitable for all lab applications.
Download
Minisart Syringe Filters
Minisart Syringe Filters
A guide to Minisart™ syringe filters suitable for multiple applications in the biotech field.
Download
Millex Nonsterile Syringe Filters
Millex Nonsterile Syringe Filters
Protect your results with Millex™ syringe filters. Choose from a broad range of membranes, pore sizes, and filter diameters to meet your needs.
Download
Shop Your Brands
Frequently Asked Questions
How do I choose the right syringe filter membrane?
This will depend on the type of solution to be filtered. For aqueous solutions, choose Nylon or MCE membranes. For non-polar solutions, choose Nylon, PTFE or PVDF membranes. For protein solutions, choose PVDF or PES membranes.
Learn More
How do I choose a suitable filter diameter?
Filter diameter is determined according to the volume of the sample. For sample volumes <10mL, choose a 13mm filter diameter. For sample volumes <100mL, choose a 25mm filter diameter. For sample volumes <250mL, choose a 33mm filter diameter.
Learn More
How do I choose a suitable pore size?
Pore size is determined according to the size (in microns) of the particles that we want to separate. For more than 3µm, choose a filter membrane with a pore size of 0.45µm and for less than 3µm, choose a poor size of 0.22µm. The lower the membrane micron rating the finer the filtration.
Learn More
Browse Other Product Categories