Learn More
Vitlab™ Wash Bottle Vent Cap
Click to view available options
Color:
Red
Yellow
Descripción
- Screw cap and spray tube
- Practically no leakage or dripping
- Design prevents almost all static overpressure
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
Especificaciones
Color | Yellow |
Descripción | Screw cap with spray tube |
Material | Polypropylene |
Tamaño del tapón | GL45 |
Tipo de tapón | Screw Cap |