Learn More
UMODL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA564317
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: CSGRELCANL EGSYWCVCHQ EAPATSPRKL NLEWEDCPPV SDYVVLNVTS DSFQVSWRLN STQNHTFHVR VYRGMELLRS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 59%.
UMODL1 Polyclonal antibody specifically detects UMODL1 in Human samples. It is validated for Immunohistochemistry.Especificaciones
UMODL1 | |
Polyclonal | |
Unconjugated | |
UMODL1 | |
D17Ertd488e; Olfactorin; RGD1566039; Umodl1; uromodulin like 1; uromodulin-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5DID0 | |
UMODL1 | |
Recombinant protein corresponding to Human UMODL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only