Learn More
Ubiquitin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532634
Detalles adicionales : Peso : 0.01000kg
Descripción
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is pancreas or Alzheimer's brain.
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by a 26S complex, multicatalytic cytosolic and nuclear protease termed proteasome. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.Especificaciones
Ubiquitin | |
Polyclonal | |
RPS27A | |
RPS27A, UBA52, UBC, polyubiquitin B, polyubiquitin-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6233, 78294, 100912032 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Unconjugated | |
P62979, P62983, P62982 | |
RPS27A | |
Recombit full length human ubiquitin protein. | |
500 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |