missing translation for 'onlineSavingsMsg'
Learn More

Tiotriazinona, +98 %, Thermo Scientific Chemicals

Catalog Number p-7026590
Click to view available options
Cantidad:
10 g
50 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
58909-39-0
C4H5N3O2S
159.163
MFCD00185756
UMWWHOXOVPIGFD-UHFFFAOYSA-N
thiotriazinone, tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione, unii-679wf9q9w1, 2-methyl-3-thioxo-1,2,4-triazinane-5,6-dione, 1,2,4-triazin-2-methyl-6-hydroxy-3-thio-5-one, 2-methyl-3-sulfanyl-1,2-dihydro-1,2,4-triazine-5,6-dione, 1,2,4-triazine-5,6-dione, tetrahydro-2-methyl-3-thioxo, acmc-20ajjm, ceftriaxone impurity c, ksc269g8j
3034640
2-metil-3-sulfanilideno-1,2,4-triazinano-5,6-diona
CN1C(=S)NC(=O)C(=O)N1
Este artículo no se puede devolver. Vea la política de devoluciones
58909-39-0
C4H5N3O2S
159.163
MFCD00185756
UMWWHOXOVPIGFD-UHFFFAOYSA-N
thiotriazinone, tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione, unii-679wf9q9w1, 2-methyl-3-thioxo-1,2,4-triazinane-5,6-dione, 1,2,4-triazin-2-methyl-6-hydroxy-3-thio-5-one, 2-methyl-3-sulfanyl-1,2-dihydro-1,2,4-triazine-5,6-dione, 1,2,4-triazine-5,6-dione, tetrahydro-2-methyl-3-thioxo, acmc-20ajjm, ceftriaxone impurity c, ksc269g8j
3034640
2-metil-3-sulfanilideno-1,2,4-triazinano-5,6-diona
CN1C(=S)NC(=O)C(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Thiotriazinone
Punto de fusión 168°C to 171°C
Cantidad 10 g
Formula Weight (peso de la fórmula) 159.16
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'