Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
This Quick Column Change accessory is compatible with Agilent Instrumentation
Marca: Thermo Scientific™ 60180-878
Detalles adicionales : Peso : 0.30000kg
Descripción
QCC (Quick Column Change)-Leak Free Column Installation
- Tool-free installation of analytical GC column
- Graphite ferrules
Complete QCC Kits Includes:
- Injector Adaptor
- Detector Adaptor
- Tight Nut
- Ferrules (1 each 0.25, 0.32,0.53)
- Ceramic Column Cutter
Especificaciones
Adapter | |
TCD Detector Adaptor |