Learn More
Thermo Scientific™ Hypersil GOLD™ C8 Javelin™ Guard Columns
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
Marca: Thermo Scientific™ 25205-013006
Detalles adicionales : Peso : 0.10000kg
Descripción
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
Features of Hypersil GOLD C8 Solid Phase
- Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
- Lower hydrophobicity
- Faster separations
- Excellent peak shape
- High efficiency
- Outstanding sensitivity
Información de órdenes
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Especificaciones
5 μm | |
10 mm | |
3 mm | |
C8 |