missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF7L Monoclonal Antibody (3E10), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524476
Detalles adicionales : Peso : 0.01000kg
Descripción
Peptide Sequence: KKRFRKTQKK VPDVKEMEKS SFTEYIESPD VENEVKRLLR SDAEAVSTRW EVIAEDGTKE IESQGSIPGF LISSGMSSHK QGHTSSEYDM LREMFSDSRS NN
TAF7L Monoclonal antibody specifically detects TAF7L in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Especificaciones
TAF7L | |
Monoclonal | |
Unconjugated | |
TAF7L | |
4933438I11Rik; 50kDa; cancer/testis antigen 40; CT40; RGD1564898; RNA polymerase II TBP-associated factor subunit Q; TAF2Q; Taf7l; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q; TATA box binding protein associated factor 7 like; TATA box binding protein-associated factor RNA polymerase II subunit Q; TATA box-binding protein-associated factor 50 kDa; TATA-box binding protein associated factor 7 like; TBP-associated factor, RNA polymerase II, Q; transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like | |
Mouse | |
Protein A | |
RUO | |
54457 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
3E10 | |
PBS with no preservative; pH 7.4 | |
Q5H9L4 | |
TAF7L | |
TAF7L (NP_079161.2, 231 a.a. ∽ 332 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |