missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudán II

Catalog Number p-7023748
Click to view available options
Cantidad:
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
3118-97-6
C18H16N2O
276.34
MFCD00003896
YPXOPAFVVHXQDP-ZZEZOPTASA-N
C.I. 12140; Solvent Orange 7
5809936
(1Z)-1-[2-(2,4-dimetilfenil)hidrazina-1-ilideno]-1,2-dihidronaftalen-2-ona
CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
3118-97-6
C18H16N2O
276.34
MFCD00003896
YPXOPAFVVHXQDP-ZZEZOPTASA-N
C.I. 12140; Solvent Orange 7
5809936
(1Z)-1-[2-(2,4-dimetilfenil)hidrazina-1-ilideno]-1,2-dihidronaftalen-2-ona
CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sudan II
Punto de fusión 156°C to 160°C
Formula Weight (peso de la fórmula) 276.34
Cantidad 25 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'