Learn More
Escamas de acero inoxidable, malla -150, 1,2 micrómetros de grosor, tipo 316-L, Thermo Scientific Chemicals
Marca: Thermo Scientific Alfa Aesar 042486.A1
Detalles adicionales : Peso : 1.00000kg
Descripción
Stainless Steel is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Stainless Steel flake, Type 316-L | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
1 kg | |
−150 Mesh |
1.2 micron thick | |
MFCD00148499 | |
Insoluble in water. | |
Escamas |
RUO – Research Use Only