missing translation for 'onlineSavingsMsg'
Learn More

Esponja de níquel, A-5000, no promocionado, Thermo Scientific Chemicals

Catalog Number p-4408748
Click to view available options
Cantidad:
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
ácido nítrico
O[N+]([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
ácido nítrico
O[N+]([O-])=O

Lodo en agua

Hydrogenation of nitro groups to amines, hydrogenation of carbonyl groups, hydrogenation of olefins to alkanes, reductive alkylation (stereoselective)Sponge Nickel, A-5000, nonpromoted is a standard catalyst used for most hydrogenation reactions such as hydrogenation of xylose to xylitol. It is also involved in the industrial hydrogenation of butyraldehyde to butanol, 2,4-dinitrotoluene to 2,4-diaminotoluene. Further, it is used as a catalyst in reductive alkylation reaction, especially tertiary diemethylaliphatic amine from aliphatic primary amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Nickel
Color Plata
Nombre de nota Nonpromoted
Aspecto Silver colored sponge
Cantidad 500 g
Información de solubilidad Insoluble in water.
Olor Odorless
Sensibilidad Air Sensitive
Formulario Lodo en agua

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'