Learn More
Novus Biologicals™ Rat MMP-14/MT1-MMP ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Marca:  Novus Biologicals™ NBP3-06939
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18349403
  • 777.52€ / 1 set
Fecha estimada de envío: 17-07-2024
para ver el stock
Especificaciones
0.16 to 10 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat MMP-14/MT1-MMP in samples. No significant cross-reactivity or interference between Rat MMP-14/MT1-MMP and analogues was observed.
4323
1 Kit
Storage of components varies. See protocol for specific instructions.
Rat
0.09 ng/mL
Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
MMP14
Cancer, Extracellular Matrix
EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1