Learn More
Novus Biologicals™ Rat MMP-14/MT1-MMP ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Marca: Novus Biologicals™ NBP3-06939
Detalles adicionales : Peso : 0.00970kg
Especificaciones
0.16 to 10 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat MMP-14/MT1-MMP in samples. No significant cross-reactivity or interference between Rat MMP-14/MT1-MMP and analogues was observed. | |
4323 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Rat |
0.09 ng/mL | |
Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
MMP14 | |
Cancer, Extracellular Matrix | |
EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 |