Learn More
Tubo para cultivo de fondo redondo de vidrio de borosilicato Pyrex™ con tapón de rosca
Ideal for culture work with PBT screw caps and PTFE lined rubber disc. PryexTM culture tubes come in a wide range of sizes to meet your needs.
Marca: Pyrex 1636/36MP
Detalles adicionales : Peso : 1.36000kg
Descripción
- Ideal for culture work
- Manufactured from Pyrex™ borosilicate glass
- High resistance to attack from water reduces leaching of contaminants which can cause pH changes
- Phenolic screw caps with inert PTFE lined rubber disc
- Cap can withstand repeated dry heat and steam sterilisation cycles (121°C for two hours)