Learn More
Propylene Glycol, U.S.P. - F.C.C., J.T. Baker™
CAS | 57-55-6 |
---|---|
Fórmula molecular | C3H8O2 |
Peso molecular (g/mol) | 76.095 |
Clave InChI | DNIAPMSPPWPWGF-UHFFFAOYSA-N |
Sinónimo | 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost |
PubChem CID | 1030 |
ChEBI | CHEBI:16997 |
Nombre IUPAC | Propano-1,2-diol |
SMILES | CC(CO)O |
57-55-6 | |
76.095 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CHEBI:16997 | |
CC(CO)O |
C3H8O2 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
1030 | |
Propano-1,2-diol |
Especificaciones
Propylene Glycol | |
C3H8O2 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
Propano-1,2-diol | |
1030 | |
76.1 | |
FCC/Farmacopea Estadounidense |
57-55-6 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CC(CO)O | |
76.095 | |
CHEBI:16997 | |
99.00°C | |
1L = 1.04kg |