missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRKACB (PRKAC beta), GST Tag Recombinant Protein

Human PRKACB (PRKAC beta), GST Tag Recombinant Protein

Marca:  Invitrogen™ PV6389

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15499013

  • 1678.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The PKA catalytic subunit consists of three gene products: C-alpha, C-beta, and C-gamma. The catalytic subunit C-beta has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Especificaciones

Especificaciones

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
66.9kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
100μg
PKAb, cAPKb, MGC9320, MGC41879, DKFZp781I2452
Recombinant
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales