missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Solution, In H2O (saturated), Honeywell™

Catalog Number 15643550
Click to view available options
Cantidad:
500 mL
Envase:
Poly bottle
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

In H2O (saturated)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Potassium Chloride Solution
CAS 7447-40-7
Color Colorless
pH 5.0 to 8.0 (at 260g/L 25°C)
Forma física Liquid
Cantidad 500 mL
Fórmula molecular ClK
Fórmula lineal KCl
Número MDL MFCD00011360
Número UN NONH for all modes of transport
Beilstein 1711999
Sinónimo potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
Clave InChI WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
Nombre IUPAC potassium chloride
Peso molecular (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight (peso de la fórmula) 74.55g/mol
Envase Poly bottle
Densidad 1.175g/mL at 25°C
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'